Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gastrokine 1 Mouse anti-Human, Rat, Clone: 3C10E3, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | Gastrokine 1 |
---|---|
Clone | 3C10E3 |
Concentration | 1.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6256
![]() ![]() |
Proteintech Group Inc
601301IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Gastrokine 1, also known as GKN1 and CA11, is a 185 amino acid containing secreted protein belonging to the gastrokine family with a BRICHOS domain and an asn-rich region. GKN1 (also known as an antioncogene) is reported to be down-regulated in human gastric cancer tissue as compared to normal gastric mucosa. Differential display of normal and cancerous gastric tissue suggested that this protein is predomitly expressed in stomach and the expression is down-regulated to considerable frequency in cancer tissue or gastric cancer cell lines examined. This extra cellular protein is co-secreted with mucins and is a signaling growth factor with mitogenic activity and may be involved in maintaining the integrity of the gastric mucosal epithelium.Specifications
Gastrokine 1 | |
1.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
18 kDa antrum mucosa protein, AMP 18, AMP18, BRICD1, CA11, FOV, foveolin, gastrokine 1, GKN1, Protein CA11 | |
Gkn1 | |
IgG2a | |
Caprylic Acid Precipitation | |
Antibody |
3C10E3 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Rat | |
Q9NS71 | |
297418, 56287 | |
Gastrokine 1 Fusion Protein Ag6091 | |
Primary | |
-20°C | |
GKN1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title