Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Gastrokine 2 Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | Gastrokine 2 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119134
|
Invitrogen™
PA5119134 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121965
|
Invitrogen™
PA5121965 |
20 μL |
Each for $209.50
|
|
|||||
Description
Gastrokine 2 Polyclonal Antibody for Western Blot, IHC (P)
The function of this protein remains unknown.Specifications
Gastrokine 2 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human, Mouse, Rat | |
GKN2 | |
1810036H07Rik; Blot; Blottin; BRICD1B; BRICHOS domain containing 1B; Down-regulated in gastric cancer; down-regulated in gastric cancer GDDR; gastrokine 2; gastrokine-2; GDDR; Gkn2; PRO813; RGD1311934; TFIZ1; trefoil factor interactions(z) 1; UNQ465/PRO813; VLTI465 | |
GKN2 | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q29TV8, Q86XP6, Q9CQS6 | |
200504, 297419, 66284 | |
Recombinant protein Gastrokine 2. The antigen corresponds to amino acid range 21-184 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title