Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GAT-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580031
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580031 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580031 Supplier Invitrogen™ Supplier No. PA580031
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Brain Tissue, Mouse Brain Tissue. IHC: Rat Cerebellum Tissue.

GAT-1 (SLC6A1) The protein encoded by SLC6A1 is a gamma-aminobutyric acid (GABA) transporter that localizes to the plasma membrane. The encoded protein removes GABA from the synaptic cleft, restoring it to presynaptic terminals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAT-1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene SLC6A1
Gene Accession No. P23978, P30531, P31648
Gene Alias A730043E01; GABA T1; GABA T-1; GABA transporter 1; GABA transporter protein; GABAT1; GABATHG; GABATR; Gabt; GABT1; gamma-aminobutyric acid (GABA-A) transporter 1; GAT1; GAT-1; MAE; SLC6A1; Sodium- and chloride-dependent GABA transporter 1; solute carrier family 6 (neurotransmitter transporter), member 1; solute carrier family 6 (neurotransmitter transporter, GABA), member 1; solute carrier family 6 member 1; XT-1; Xtrp1
Gene Symbols SLC6A1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human GABA Transporter 1 (473-487aa WFYGVNRFYDNIQEM).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 232333, 6529, 79212
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.