Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GAT-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590220
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590220 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590220 Supplier Invitrogen™ Supplier No. PA590220
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse eye, Mouse brain, Rat brain; Cellular Location: Cell membrane, Membrane, Multi-pass membrane protein Immunogen sequence: WSFFTPIIVA GVFIFSAVQM TPLTMGNYVF PKWGQGVGWL MALSSMVLIP GYMAYMFLTL KGSLKQRIQV MVQPSEDIVR PENGPEQPQA GSSTSKEAYI.

GAT-1 (SLC6A1) The protein encoded by SLC6A1 is a gamma-aminobutyric acid (GABA) transporter that localizes to the plasma membrane. The encoded protein removes GABA from the synaptic cleft, restoring it to presynaptic terminals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAT-1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.11 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SLC6A1
Gene Accession No. P23978, P30531, P31648
Gene Alias A730043E01; GABA T1; GABA T-1; GABA transporter 1; GABA transporter protein; GABAT1; GABATHG; GABATR; Gabt; GABT1; gamma-aminobutyric acid (GABA-A) transporter 1; GAT1; GAT-1; MAE; SLC6A1; Sodium- and chloride-dependent GABA transporter 1; solute carrier family 6 (neurotransmitter transporter), member 1; solute carrier family 6 (neurotransmitter transporter, GABA), member 1; solute carrier family 6 member 1; XT-1; Xtrp1
Gene Symbols SLC6A1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 500-599 of human SLC6A1 (NP_003033.3).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 232333, 6529, 79212
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.