Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAT-1/SLC6A1 Antibody (SR2118), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32197050UL
Description
GAT-1/SLC6A1 Monoclonal antibody specifically detects GAT-1/SLC6A1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
GAT-1/SLC6A1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
SR2118 | |
Western Blot 1:500-1:2000 | |
GABA Transporter 1, GABATHG, GABATRGAT-1, GABT1, GAT1sodium- and chloride-dependent GABA transporter 1, solute carrier family 6 (neurotransmitter transporter, GABA), member 1, Solute carrier family 6 member 1 | |
A synthesized peptide derived from human GAT-1/SLC6A1 (Uniprot #: P30531) | |
50 μg | |
Neuroscience | |
6529 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction