Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GATA-2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 6 publications
Supplier: Novus Biologicals NBP182581
Description
GATA-2 Polyclonal specifically detects GATA-2 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
GATA-2 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
endothelial transcription factor GATA-2, FLJ45948, GATA binding protein 2, GATA-binding protein 2NFE1B, MGC2306 | |
Rabbit | |
51 kDa | |
0.1 mL | |
Primary | |
Specificity of human GATA-2 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
0.6mg/mL | |
Western Blot 0.04-0.4 μg/mL, Simple Western 1:100, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 0.25 - 2 μg/mL, Immunoprecipitation, Immunohistochemistry-Paraffin 1:20 - 1:50, Immunohistochemistry-Frozen 1:200 - 1:500 | |
P23769 | |
GATA2 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:LTGGQMCRPHLLHSPGLPWLDGGKAALSAAAAHHHNPWTVSPFSKTPLHPSAAGGPGGPLSVYPGAGGGSGGGSGSSVASLTPTAAHSGSHLFGFPPTPPKEVSPDPSTTGAASPASSSAGGSAARG | |
Affinity Purified | |
RUO | |
2624 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction