Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GATA-3 Antibody (634919), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB26052A
Description
GATA-3 Monoclonal antibody specifically detects GATA-3 in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
GATA-3 | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry | |
634919 | |
Western Blot, Intracellular Staining by Flow Cytometry | |
131320, GATA binding protein 3, GATA-binding factor 3, GATA-binding protein 3, HDRMGC5199, MGC2346, MGC5445, trans-acting T-cell-specific transcription factor GATA-3 | |
E. coli-derived recombinant human GATA-3, Pro135-Ser258, Accession # P23771 | |
0.1 mL | |
Cancer, Innate Immunity | |
2625 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction