Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GATA-3 Antibody (GATA3/4550R), HRP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP314267H
This item is not returnable.
View return policy
Description
GATA-3 Monoclonal antibody specifically detects GATA-3 in Human, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
GATA-3 | |
Monoclonal | |
HRP | |
PBS | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Rat | |
Antibody | |
IgG κ |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
GATA3/4550R | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
131320, GATA binding protein 3, GATA-binding factor 3, GATA-binding protein 3, HDRMGC5199, MGC2346, MGC5445, trans-acting T-cell-specific transcription factor GATA-3 | |
Recombinant full-length human GATA-3 protein (Uniprot: P23771) | |
0.1 mL | |
Cancer, Innate Immunity | |
2625 | |
Store at 4°C in the dark. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction