Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATA-3 Mouse anti-Human, Clone: GATA3/2688, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00

Specifications

Antigen GATA-3
Clone GATA3/2688
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA, Protein Array
Applications Western Blot, Flow Cytometry, ELISA, Peptide Array
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122884
SDP
View Documents
Novus Biologicals
NBP30770520UG
20 μg
Each of 1 for $268.00
Only null left
Add to Cart
 
Description

Description

GATA-3 Monoclonal specifically detects GATA-3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Protein Array.
Specifications

Specifications

GATA-3
0.2 mg/ml
Western Blot, Flow Cytometry, ELISA, Peptide Array
Unconjugated
Mouse
Cancer, Innate Immunity
10 mM PBS with 0.05% BSA
2625
IgG1 κ
Protein A or G purified
GATA3/2688
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA, Protein Array
Monoclonal
Purified
RUO
Human
131320, GATA binding protein 3, GATA-binding factor 3, GATA-binding protein 3, HDRMGC5199, MGC2346, MGC5445, trans-acting T-cell-specific transcription factor GATA-3
Recombinant fragment of human GATA-3 protein (around aa 357-436) (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.