Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GATA-4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP256566
Description
GATA-4 Polyclonal specifically detects GATA-4 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
GATA-4 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
GATA binding protein 4, GATA-binding factor 4, GATA-binding protein 4, MGC126629, transcription factor GATA-4 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
GATA4 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PFDSPVLHSLPGRANPAARHPNLDMFDDFSEGRECV | |
100 μL | |
Mesenchymal Stem Cell Markers, Stem Cell Markers | |
2626 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction