Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GATA-4 Antibody (OTI9F9), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270797FR
Description
GATA-4 Monoclonal specifically detects GATA-4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
GATA-4 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
GATA4 | |
Recombinant protein fragment corresponding to amino acids 296-442 of human GATA-4 (NP_002043) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG3 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI9F9 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
GATA binding protein 4, GATA-binding factor 4, GATA-binding protein 4, MGC126629, transcription factor GATA-4 | |
Mouse | |
Affinity Purified | |
Mesenchymal Stem Cell Markers, Phospho Specific, Stem Cell Markers | |
2626.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction