Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GATA-6 Polyclonal Antibody, Active Motif™

Rabbit Polyclonal Antibody
$156.65 - $620.58
Specifications
Antigen | GATA-6 |
---|---|
Concentration | Lot-Specific |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3103
![]() ![]() |
Active Motif
61064 |
10 μL |
Each for $156.65
|
|
|||||
50-199-3102
![]() ![]() |
Active Motif
61063 |
100 μL |
Each for $620.58
|
|
|||||
Description
GATA6 is thought to be important for regulating terminal differentiation and/or proliferation.Specifications
GATA-6 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q92908 | |
2627 | |
This GATA-6 antibody was raised against a peptide within the N-terminus of human GATA-6. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
GATA6 |
Lot-Specific | |
Polyclonal | |
Liquid | |
RUO | |
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8 | |
AA410133; ASD9; AVSD5; BOS_22226; DNA-binding protein GATA-GT2; GATA binding protein 6; Gata6; GATA-6; GATA-binding factor 6; GATA-binding protein 6; LOW QUALITY PROTEIN: transcription factor GATA-6; transcription factor GATA6; Transcription factor GATA-6 | |
GATA6 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title