Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GATA1 Polyclonal Antibody

Catalog No. PIPA1099X
Change view
Click to view available options
Quantity:
100 μg
20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA1099X 20 μL
PIPA1099 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA1099X Supplier Invitrogen™ Supplier No. PA1099X
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-099 specifically detects GATA1 and does not cross-react with GATA2 or GATA3.

Transcriptional activator or repressor which probably serves as a general switch factor for erythroid development. It binds to DNA sites with the consensus sequence 5'-[AT]GATA[AG]-3' within regulatory regions of globin genes and of other genes expressed in erythroid cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GATA1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide
Gene Gata1
Gene Accession No. P15976, P17679
Gene Alias eryf1; Erythroid transcription factor; erythroid transcription factor 1; GATA binding protein 1; GATA binding protein 1 (globin transcription factor 1); GATA1; GATA-1; GATA-binding factor 1; GATA-binding protein 1; GATA-binding protein 1 (globin transcription factor 1); GF1; Gf-1; globin transcription factor 1; HGNC:4170; NFE1; NF-E1; NF-E1 DNA-binding protein; nuclear factor, erythroid 1; transcription factor GATA1; XLANP; XLTDA; XLTT
Gene Symbols Gata1
Host Species Rabbit
Immunogen N-terminal truncated human recombinant protein.
Purification Method Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14460, 2623
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.