Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gata1 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501992295
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2295 100 μg
50-199-2296 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2295 Supplier Rockland Immunochemicals Supplier No. 600401K12
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-Gata1 is directed against mouse Gata1. A BLAST analysis was used to suggest reactivity with this protein from mouse and rat based on 100% homology for the immunogen sequence and 93% to human. Cross reactivity with Gata1 homologues from other sources has not been determined.

Transcriptional activator or repressor which probably serves as a general switch factor for erythroid development. It binds to DNA sites with the consensus sequence 5'-[AT]GATA[AG]-3' within regulatory regions of globin genes and of other genes expressed in erythroid cells.

Specifications

Antigen Gata1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.73 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene Gata1
Gene Accession No. P17679
Gene Alias ERYF1; Erythroid transcription factor; erythroid transcription factor 1; GATA binding protein 1; GATA binding protein 1 (globin transcription factor 1); GATA1; GATA-1; GATA-binding factor 1; GATA-binding protein 1; GATA-binding protein 1 (globin transcription factor 1); GF1; Gf-1; globin transcription factor 1; HGNC:4170; NFE1; NF-E1; NF-E1 DNA-binding protein; nuclear factor, erythroid 1; transcription factor GATA1; XLANP; XLTDA; XLTT
Gene Symbols Gata1
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14460
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.