Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GATA2/GATA3 Recombinant Rabbit Monoclonal Antibody (1K8H6)
GREENER_CHOICE

Catalog No. PIMA535637
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535637 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535637 Supplier Invitrogen™ Supplier No. MA535637
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: MEVAPEQPRW MAHPAVLNAQ HPDSHHPGLA HNYMEPAQLL PPDEVDVFFN HLDSQGNPYY ANPAHARARV SYSPAHARLT GGQMCRPHLL HSPGLPWLDG GKAALSAAAA HHHNPWTVSP FSKTPLHPSA AGGPGGPLSV YPGAGGGSGG GSGSSVASLT PTAAHSGSHL FGFPPTPPKE VSPDPSTTGA ASPASSSAGG SAARGEDKDG.

The genes for all 4 subunits of the T-cell antigen receptor (alpha, beta, gamma and delta) are controlled by distinct enhancers and their enhancer-binding proteins. Marine and Winoto (1991) identified a common TCR regulatory element by demonstrating binding of the enhancer-binding protein GATA3 to the enhancer elements of all 4 TCR genes. GATA3 had been shown in the chicken to be an enhancer-binding protein containing a zinc finger domain. GATA3 mRNA was demonstrated by Northern blot analysis in T cells but not in B cells or macrophages. GATA3 is abundantly expressed in the T-lymphocyte lineage and is thought to participate in T-cell receptor gene activation through binding to enhancers. Labastie et al. (1994) cloned the human gene and the 5-prime end of the mouse gene. The human gene comprises 6 exons distributed over 17 kb of DNA. Its 2 zinc fingers are encoded by 2 separate exons highly conserved with those of GATA1, but no other structural homologies between the 2 genes could be found.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GATA2/GATA3
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 1K8H6
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GATA2
Gene Accession No. O09100, P23769, P23771, P23772, Q924Y4
Gene Alias DCML; EMBL:AAK51128.1}; endothelial transcription factor GATA-2; GATA binding protein 2; GATA binding protein 3; Gata2; Gata-2; gata2 {ECO:0000312; Gata3; Gata-3; GATA-binding factor 3; GATA-binding protein 2; GATA-binding protein 3; GATA-bindning protein 2; HDR; HDRS; IMD21; jal; MGC2346; MGC5199; MGC5445; MONOMAC; NFE1B; trans-acting T-cell-specific transcription factor GATA-3
Gene Symbols GATA2, GATA3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human GATA2/3 (NP_1160272).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14461, 14462, 25159, 2624, 2625, 85471
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.