Learn More
Invitrogen™ GATA2/GATA3 Polyclonal Antibody

Description
Antibody detects endogenous levels of total GATA2/3.
Specifications
Specifications
| Antigen | GATA2/GATA3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | GATA2 |
| Gene Accession No. | O09100, P23769, P23771, P23772, Q924Y4 |
| Gene Alias | DCML; EMBL:AAK51128.1}; endothelial transcription factor GATA-2; GATA binding protein 2; GATA binding protein 3; Gata2; Gata-2; gata2 {ECO:0000312; Gata3; Gata-3; GATA-binding factor 3; GATA-binding protein 2; GATA-binding protein 3; GATA-bindning protein 2; HDR; HDRS; IMD21; jal; MGC2346; MGC5199; MGC5445; MONOMAC; NFE1B; trans-acting T-cell-specific transcription factor GATA-3 |
| Gene Symbols | GATA2, GATA3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.