Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATA2 Mouse anti-Human, Unconjugated, Clone: 3C10.1, MilliporeSigma™

Catalog No. MABS181425 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS1814-25 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1814-25 Supplier MilliporeSigma Supplier No. MABS181425UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-GATA2 Antibody, clone 3C10.1, Cat. No. MABS1814, is a mouse monoclonal antibody that targets GATA2 and has been tested in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GATA2
Applications Western Blot
Classification Monoclonal
Clone 3C10.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Endothelial transcription factor GATA-2;GATA-binding protein 2
Gene Symbols GATA2
Host Species Mouse
Immunogen GST/His-tagged recombinant fragment corresponding to 114 amino acids from the internal region of human GATA2.
Molecular Weight of Antigen ∽52 kDa observed; 50.50 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity Clone 3C10.1 specifically detects Endothelial transcription factor GATA-2 in human. It targets an epitope within 114 amino acids from the internal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.