Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GATA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA1100
Change view
Click to view available options
Quantity:
100 μg
20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA1100 100 μg
PIPA1100X 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA1100 Supplier Invitrogen™ Supplier No. PA1100
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-100 specifically detects GATA2 and does not cross-react with GATA1 or GATA3.

GATA2 (GATA binding protein 2) belongs to the GATA family of transcription factors. GATA proteins play a very important role in signaling pathways controlling proliferation and differentiation of erythroid progenitor cells. There are 6 transcription factors in the GATA family: GATA1 to GATA6. GATA2 is a transcriptional activator regulating endothelin-1 gene expression in endothelial cells. It also appears to be involved in differentiation of other tissue types (uro-genital and nervous) during embryogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GATA2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide
Gene GATA2
Gene Accession No. O09100, P23769
Gene Alias DCML; EMBL:AAK51128.1}; Endothelial transcription factor GATA-2; FLJ45948; GATA binding protein 2; GATA binding protein 3; GATA2; Gata-2; gata2 {ECO:0000312; GATA3; GATA-binding factor 3; GATA-binding protein 2; GATA-bindning protein 2; HDR; HDRS; IMD21; MGC2306; MONOMAC; NFE1B; trans-acting T-cell-specific transcription factor GATA-3; uterine GATA-4 transcription factor
Gene Symbols GATA2
Host Species Rabbit
Immunogen N-terminal truncated human recombinant protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14461, 2624
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.