Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GATA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529736
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529736 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529736 Supplier Invitrogen™ Supplier No. PA529736
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HeLa, HepG2, HCT116, H1299. Predicted reactivity: Mouse (97%), Rat (96%), Xenopus laevis (87%), Pig (99%), Chicken (86%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

GATA2 (GATA binding protein 2) belongs to the GATA family of transcription factors. GATA proteins play a very important role in signaling pathways controlling proliferation and differentiation of erythroid progenitor cells. There are 6 transcription factors in the GATA family: GATA1 to GATA6. GATA2 is a transcriptional activator regulating endothelin-1 gene expression in endothelial cells. It also appears to be involved in differentiation of other tissue types (uro-genital and nervous) during embryogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GATA2
Applications Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene GATA2
Gene Accession No. O09100, P23769, Q924Y4
Gene Alias DCML; EMBL:AAK51128.1}; Endothelial transcription factor GATA-2; FLJ45948; GATA binding protein 2; GATA binding protein 3; GATA2; Gata-2; gata2 {ECO:0000312; GATA3; GATA-binding factor 3; GATA-binding protein 2; GATA-bindning protein 2; HDR; HDRS; IMD21; MGC2306; MONOMAC; NFE1B; trans-acting T-cell-specific transcription factor GATA-3; uterine GATA-4 transcription factor
Gene Symbols GATA2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human GATA2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14461, 25159, 2624
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.