Learn More
Invitrogen™ GATA3 Monoclonal Antibody (7B5)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA515641
Description
MA5-15641 targets GATA3 in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15641 immunogen is purified recombinant fragment of human GATA3 expressed in E. Coli. . MA5-15641 detects GATA3 which has a predicted molecular weight of approximately 48kDa.
The genes for all 4 subunits of the T-cell antigen receptor (alpha, beta, gamma and delta) are controlled by distinct enhancers and their enhancer-binding proteins. Marine and Winoto (1991) identified a common TCR regulatory element by demonstrating binding of the enhancer-binding protein GATA3 to the enhancer elements of all 4 TCR genes. GATA3 had been shown in the chicken to be an enhancer-binding protein containing a zinc finger domain. GATA3 mRNA was demonstrated by Northern blot analysis in T cells but not in B cells or macrophages. GATA3 is abundantly expressed in the T-lymphocyte lineage and is thought to participate in T-cell receptor gene activation through binding to enhancers. Labastie et al. (1994) cloned the human gene and the 5-prime end of the mouse gene. The human gene comprises 6 exons distributed over 17 kb of DNA. Its 2 zinc fingers are encoded by 2 separate exons highly conserved with those of GATA1, but no other structural homologies between the 2 genes could be found.
Specifications
GATA3 | |
Monoclonal | |
Conc. Not Determined | |
Ascites with 0.03% sodium azide | |
P23771 | |
GATA3 | |
Purified recombinant fragment of human GATA3 expressed in E. Coli. | |
RUO | |
2625 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
7B5 | |
Unconjugated | |
GATA3 | |
EGK_19445; GATA binding factor-3; GATA binding protein 3; GATA binding protein-3; GATA3; Gata-3; GATA-binding factor 3; GATA-binding protein 3; HDR; HDRS; jal; MGC2346; MGC5199; MGC5445; Trans-acting T-cell-specific transcription factor GATA-3; Transacting T-cell-specific transcription factor GATA-3 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.