Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATA3 Mouse, Alexa Fluor 488, Clone: L50-823, BD

Catalog No. BDB560163 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
50 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560163 50 Tests
BDB560077 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB560163 Supplier BD Biosciences Supplier No. 560163
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GATA3 (GATA binding protein 3) is a member of the GATA family of transcription factors. This ∽50kDa nuclear protein regulates the development and subsequent maintenance of multiple tissues. GATA3 is involved in the development of T lymphocytes (regulates T cell receptor subunit gene expression) and the differentiation of mature T cells to become Th2 cells. The expressed levels of normal or mutant GATA3 are also associated with the behaviors of various cancer cells including estrogen receptor-positive breast carcinoma cells. The L50-823 monoclonal antibody recognizes human and mouse GATA3.

Host Species: Mouse
Clone: L50-823
Isotype: IgG1 κ
Species Reactivity: Human, Mouse
Immunogen: Conserved peptide between the trans-activation and DNA-binding domains of human, mouse and rat GATA3

Bioimaging

TRUSTED_SUSTAINABILITY

Specifications

Antigen GATA3
Applications Flow Cytometry
Classification Monoclonal
Clone L50-823
Conjugate Alexa Fluor 488
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Conserved peptide between the trans-activation and DNA-binding domains of human, mouse and rat GATA3
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.