Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATA3 Mouse anti-Human, Mouse, Rat, Clone: 1A12-1D9, Abnova™

Catalog No. 89035470 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-470 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-470 Supplier Abnova Corporation Supplier No. MAB14065

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against recombinant human GATA3.

  • High specificity
  • High reactivity

Specifications

Antigen GATA3
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1A12-1D9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant human GATA3.
Dilution Immunocytochemistry (1:50-1:100) Immunofluorescence (1:50-1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:2000) Immunoprecipitation (5 ug) Western Blot (1:1000) The optimal working dilution should be det
Formulation In PBS (30% glycerol, 1 mg/mL BSA, 0.05% sodium azide).
Gene Accession No. P23771
Gene Alias HDR/MGC2346/MGC5199/MGC5445
Gene Symbols GATA3
Host Species Mouse
Immunogen Recombinant protein corresponding to human GATA3.
Purification Method Protein A affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2625
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.