Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATA3 Mouse anti-Human, Unconjugated, Clone: GATA3/2688, Abnova™

Catalog No. 89337503 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-503 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-503 Supplier Abnova Corporation Supplier No. MAB21010
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

GATA3 Monoclonal antibody specifically detects GATA3 in Human samples. It is validated for IHC-P.

Specifications

Antigen GATA3
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone GATA3/2688
Conjugate Unconjugated
Description GATA binding protein 3
Gene GATA3
Gene Alias HDR, MGC2346, MGC5199, MGC5445
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 357-436 of human GATA3
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 2625
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.