Learn More
Description
Specifications
Specifications
| Antigen | GATA3 |
| Applications | Flow Cytometry, Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Clone | L50-823 |
| Conjugate | Brilliant Ultraviolet 395 |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Gene Alias | Gata-3; GATA binding protein 3; GATA-binding factor 3; HDR |
| Host Species | Mouse |
| Immunogen | Conserved peptide between the trans-activation and DNA-binding domains of human, mouse and rat GATA3 |
| Quantity | 50 Tests |
| Show More |
For Reseach Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.