Learn More
Invitrogen™ GATA3 Polyclonal Antibody

Description
A suggested positive control is human brain tissue lysate. PA5-20892 can be used with blocking peptide PEP-1006.
Specifications
Specifications
| Antigen | GATA3 |
| Applications | ChIP Assay, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | GATA3 |
| Gene Accession No. | P23771, P23772 |
| Gene Alias | EGK_19445; GATA binding factor-3; GATA binding protein 3; GATA binding protein-3; GATA3; Gata-3; GATA-binding factor 3; GATA-binding protein 3; HDR; HDRS; jal; MGC2346; MGC5199; MGC5445; Trans-acting T-cell-specific transcription factor GATA-3; Transacting T-cell-specific transcription factor GATA-3 |
| Gene Symbols | GATA3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.