Learn More
GATA4 Monoclonal Antibody (10D8.B10.D4), Rockland™

Mouse Monoclonal Antibody
$178.35 - $665.43
Specifications
| Antigen | GATA4 |
|---|---|
| Clone | 10D8.B10.D4 |
| Concentration | 1 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-0670
![]()
|
Rockland Immunochemicals
200301P15 |
100 μg |
Each for $665.43
|
|
|||||
50-199-0671
![]()
|
Rockland Immunochemicals
200301P15S |
25 μL |
Each for $178.35
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. A BLAST analysis was used to suggest reactivity with mouse and rat GATA4 based on a 93% homology.Cross-reactivity with GATA4 from other sources has not been determined.
GATA4 is a member of the GATA family of zinc-finger transcription factors. Members of this family recognize the GATA motif which is present in the promoters of many genes. This protein is thought to regulate genes involved in embryogenesis and in myocardial differentiation and function. Mutations in the gene encoding GATA4 have been associated with cardiac septal defects.Specifications
| GATA4 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
| ASD2; DNA-binding protein GATA-GT2; GAT4; GATA binding protein 4; GATA4; Gata-4; GATA-binding factor 4; GATA-binding protein 4; MGC126629; TACHD; TOF; Transcription factor GATA-4; VSD1 | |
| GATA4 | |
| IgG1 κ | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| GATA4 |
| 10D8.B10.D4 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P43694 | |
| 2626 | |
| GATA4 protein A purified antibody was prepared in mice by repeated immunizations with a recombinant protein corresponding to the first 100 amino acids of human GATA4 protein. | |
| Primary | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.