Learn More
Invitrogen™ GATA4 Monoclonal Antibody (6H10)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA515532
Description
MA5-15532 targets GATA4 in indirect ELISA, WB and ChIP applications and shows reactivity with Human and mouse samples. The MA5-15532 immunogen is purified recombinant fragment of human GATA4 expressed in E. Coli. MA5-15532 detects GATA4 which has a predicted molecular weight of approximately 46kDa.
GATA4 is a member of the GATA family of zinc-finger transcription factors. Members of this family recognize the GATA motif which is present in the promoters of many genes. This protein is thought to regulate genes involved in embryogenesis and in myocardial differentiation and function. Mutations in the gene encoding GATA4 have been associated with cardiac septal defects.
Specifications
GATA4 | |
Monoclonal | |
Conc. Not Determined | |
Ascites with 0.03% sodium azide | |
P43694, P46152, Q08369 | |
GATA4 | |
Purified recombinant fragment of human GATA4 expressed in E. Coli. | |
RUO | |
14463, 2626, 54254 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ChIP Assay, ELISA, Western Blot | |
6H10 | |
Unconjugated | |
GATA4 | |
ASD2; DNA-binding protein GATA-GT2; GAT4; GATA binding protein 4; GATA4; Gata-4; GATA-binding factor 4; GATA-binding protein 4; MGC126629; TACHD; TOF; Transcription factor GATA-4; VSD1 | |
Mouse | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.