Learn More
GATA4 Mouse anti-Human, Clone: OTI9F9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500091

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
GATA4 is a member of the GATA family of zinc-finger transcription factors. Members of this family recognize the GATA motif which is present in the promoters of many genes. This protein is thought to regulate genes involved in embryogenesis and in myocardial differentiation and function. Mutations in the gene encoding GATA4 have been associated with cardiac septal defects.Specifications
GATA4 | |
Monoclonal | |
Unconjugated | |
GATA4 | |
ASD2, TACHD, TOF, VSD1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
2626 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI9F9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P43694 | |
GATA4 | |
Human recombit protein fragment corresponding to amino acids 296-442 of human GATA4 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG3 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.