Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GATA4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA511438
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA511438 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA511438 Supplier Invitrogen™ Supplier No. PA511438
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with mouse, rat and Xenopus based on sequence homology.

GATA4 is a member of the GATA family of zinc-finger transcription factors. Members of this family recognize the GATA motif which is present in the promoters of many genes. This protein is thought to regulate genes involved in embryogenesis and in myocardial differentiation and function. Mutations in the gene encoding GATA4 have been associated with cardiac septal defects.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GATA4
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 2.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene GATA4
Gene Accession No. P43694
Gene Alias ASD2; DNA-binding protein GATA-GT2; GAT4; GATA binding protein 4; GATA4; Gata-4; GATA-binding factor 4; GATA-binding protein 4; MGC126629; TACHD; TOF; Transcription factor GATA-4; VSD1
Gene Symbols GATA4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 298-328 amino acids from the C-terminal region of human GATA4.
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2626
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.