Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GATA4 Recombinant Rabbit Monoclonal Antibody (1M2E4)
GREENER_CHOICE

Catalog No. PIMA538112
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538112 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538112 Supplier Invitrogen™ Supplier No. MA538112
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: HepG2, SKOV3, Mouse liver, Mouse testis, Rat testis Immunogen sequence: ASGASSNSSN ATTSSSEEMR PIKTEPGLSS HYGHSSSVSQ TFSVSAMSGH GPSIHPVLSA LKLSPQGYAS PVSQSPQTSS KQDSWNSLVL ADSHGDIITA.

GATA4 is a member of the GATA family of zinc-finger transcription factors. Members of this family recognize the GATA motif which is present in the promoters of many genes. This protein is thought to regulate genes involved in embryogenesis and in myocardial differentiation and function. Mutations in the gene encoding GATA4 have been associated with cardiac septal defects.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GATA4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 1M2E4
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.05% BSA and 0.05% ProClin 300; pH 7.3
Gene GATA4
Gene Accession No. P46152, Q08369
Gene Alias ASD2; DNA-binding protein GATA-GT2; GAT4; GATA binding protein 4; GATA4; Gata-4; GATA-binding factor 4; GATA-binding protein 4; MGC126629; TACHD; TOF; Transcription factor GATA-4; VSD1
Gene Symbols GATA4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 343-442 of human GATA4 (NP_0020432).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14463, 54254
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.