Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GATA5 Monoclonal Antibody (1B11)
GREENER_CHOICE

Catalog No. PIMA515897
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515897 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515897 Supplier Invitrogen™ Supplier No. MA515897
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15897 targets GATA5 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15897 immunogen is purified recombinant fragment of human GATA5 expressed in E. Coli. . MA5-15897 detects GATA5 which has a predicted molecular weight of approximately 41kDa.

GATA binding factor 5 is a transcription regulator involved in embryonic development. In addition, the protein encoded by this gene contains two GATA-type zinc fingers and is known to bind to hepatocyte nuclear factor-1alpha (HNF-1alpha). This interaction is essential for cooperative activation of the intestinal lactase-phlorizin hydrolase promoter. GATA-5 may be involved in the establishment of cardiac smooth muscle cell diversity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GATA5
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1B11
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene Gata5
Gene Accession No. Q9BWX5
Gene Alias bB379O24.1; GATA binding factor-5; GATA binding protein 5; GATA5; GATA-binding factor 5; GATA-binding protein 5; GATAS; hypothetical protein LOC504784; transcription factor GATA-5; Transcription factor GATA-5 (GATA binding factor-5)
Gene Symbols Gata5
Host Species Mouse
Immunogen Purified recombinant fragment of human GATA5 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140628
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.