Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GATA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568946
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568946 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568946 Supplier Invitrogen™ Supplier No. PA568946
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Dog: 79%; Mouse: 100%; Rat: 93%.

GATA binding factor 5 is a transcription regulator involved in embryonic development. In addition, the protein encoded by this gene contains two GATA-type zinc fingers and is known to bind to hepatocyte nuclear factor-1alpha (HNF-1alpha). This interaction is essential for cooperative activation of the intestinal lactase-phlorizin hydrolase promoter. GATA-5 may be involved in the establishment of cardiac smooth muscle cell diversity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GATA5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Gata5
Gene Accession No. P97489
Gene Alias bB379O24.1; GATA binding factor-5; GATA binding protein 5; GATA5; GATA-binding factor 5; GATA-binding protein 5; GATAS; hypothetical protein LOC504784; transcription factor GATA-5; Transcription factor GATA-5 (GATA binding factor-5)
Gene Symbols Gata5
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse GATA5 (aa 1-50).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14464
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.