Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATA5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501736156 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6156 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6156 Supplier Proteintech Group Inc Supplier No. 554331AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GATA binding factor 5 is a transcription regulator involved in embryonic development. In addition, the protein encoded by this gene contains two GATA-type zinc fingers and is known to bind to hepatocyte nuclear factor-1alpha (HNF-1alpha). This interaction is essential for cooperative activation of the intestinal lactase-phlorizin hydrolase promoter. GATA-5 may be involved in the establishment of cardiac smooth muscle cell diversity.

Specifications

Antigen GATA5
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.69 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GATA5
Gene Accession No. P97489, Q9BWX5
Gene Alias bB379O24.1, GATA binding factor 5, GATA binding protein 5, GATA5, Transcription factor GATA 5
Gene Symbols GATA5
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140628, 14464
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.