Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GATA6 Polyclonal Antibody
Rabbit Polyclonal Antibody
$226.00 - $585.00
Specifications
Antigen | GATA6 |
---|---|
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Description
GATA6 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
GATA6 is thought to be important for regulating terminal differentiation and/or proliferation.Specifications
GATA6 | |
-20°C | |
Polyclonal | |
Liquid | |
IgG | |
Human, Mouse | |
GATA6 | |
AA410133; ASD9; AVSD5; DNA-binding protein GATA-GT2; GATA binding protein 6; Gata6; GATA-6; GATA-binding factor 6; GATA-binding protein 6; LOW QUALITY PROTEIN: transcription factor GATA-6; transcription factor GATA6; transcription factor GATA-6; zinc-finger transcription factor | |
GATA6 | |
Primary | |
Antigen affinity chromatography |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide | |
Q61169, Q92908 | |
14465, 2627 | |
Synthetic peptide sequence KYSGQDGLYIGVSLASPAE & KRFGAAGADASDSRAFPARE. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title