Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GATA6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540559
Description
Peptide sequence: PEEMYQTLAA LSSQGPAAYD GAPGGFVHSA AAAAAAAAAA SSPVYVPTTR Sequence homology: Cow: 92%; Dog: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%.
GATA6 is thought to be important for regulating terminal differentiation and/or proliferation.
Specifications
| GATA6 | |
| Polyclonal | |
| Unconjugated | |
| GATA6 | |
| AA410133; ASD9; AVSD5; DNA-binding protein GATA-GT2; GATA binding protein 6; Gata6; GATA-6; GATA-binding factor 6; GATA-binding protein 6; LOW QUALITY PROTEIN: transcription factor GATA-6; transcription factor GATA6; transcription factor GATA-6; zinc-finger transcription factor | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 2627 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q92908 | |
| GATA6 | |
| Synthetic peptide directed towards the N-terminal of human GATA6 (aa 144-193). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction