Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GATAD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568585
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.
Component of some chromatin complex recruited to chromatin sites methylated 'Lys-4' of histone H3 (H3K4me).
Specifications
GATAD1 | |
Polyclonal | |
Unconjugated | |
GATAD1 | |
2310031E19Rik; 2810047M21Rik; 8430439A17Rik; 9130430G15Rik; B330017N08Rik; CMD2B; ENSMUSG00000073241; GATA zinc finger domain containing 1; GATA zinc finger domain-containing 1-like protein; GATA zinc finger domain-containing protein 1; GATA zinc finger domain-containing protein 1-like protein; Gatad1; im:7150682; ocular development associated; ocular development-associated gene protein; ODAG; RG083M05.2; RG83M5.2; si:dkey-231l1.7; Unknown (protein for MGC:136582); zgc:136582 | |
Rabbit | |
Affinity chromatography | |
RUO | |
57798 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8WUU5 | |
GATAD1 | |
synthetic peptide directed towards the C terminal region of human GATAD1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction