Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GATC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570620
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570620 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570620 Supplier Invitrogen™ Supplier No. PA570620
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Human: 100%; Pig: 92%; Rabbit: 83%.

GATC (glutamyl-tRNA(Gln) amidotransferase, subunit C) is a protein-coding gene. GO annotations related to this gene include glutaminyl-tRNA synthase (glutamine-hydrolyzing) activity. An important paralog of this gene is ENSG00000111780.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GATC
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GATC
Gene Accession No. O43716
Gene Alias 15E1.2; 2010003O18Rik; Gatc; gatC-like protein; glu-AdT subunit C; glu-AdT subunit C {ECO:0000255; glutamyl-tRNA amidotransferase subunit C; glutamyl-tRNA(Gln) amidotransferase subunit C, mitochondrial; glutamyl-tRNA(Gln) amidotransferase subunit C, mitochondrial {ECO:0000255; glutamyl-tRNA(Gln) amidotransferase, subunit C; glutamyl-tRNA(Gln) amidotransferase, subunit C homolog; HAMAP-Rule:MF_03149}; Protein 15E1.2; RGD1309698; similar to CG33084-PB, isoform B
Gene Symbols GATC
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human GATC (aa 9-58).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283459
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.