Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GATC Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30444020UL
Description
GATC Polyclonal specifically detects GATC in Mouse, Rat samples. It is validated for Western Blot.Specifications
GATC | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
283459 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
FLJ37000, gatC, glutamyl-tRNA(Gln) amidotransferase, subunit C homolog (bacterial), MGC129938,15E1.2gatC-like protein, Protein 15E1.2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-136 of human GATC (NP_789788.1). | |
20 μL | |
Primary | |
Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction