Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GATD3A Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $524.00
Specifications
Antigen | GATD3A |
---|---|
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
GATD3A Polyclonal specifically detects GATD3A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
GATD3A | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
8209 | |
IgG | |
Affinity purified |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
chromosome 21 open reading frame 33, D21S2048E, ES1, GT335, HES1, HES1KNP-Ia, human HES1 protein, homolog to E.coli and zebrafish ES1 protein, 10Protein GT335, Keio novel protein I, KNPH, KNPI, KNP-I, KNPImitochondrial, Protein KNP-I | |
Recombinant fusion protein containing a sequence corresponding to amino acids 42-268 of human GATD3A (NP_004640.4). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title