Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GATM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109756
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109756 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109756 Supplier Invitrogen™ Supplier No. PA5109756
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: STQAATASSR NSCAADDKAT EPLPKDCPVS SYNEWDPLEE VIVGRAENAC VPPFTIEVKA NTYEKYWPFY QKQGGHYFPK DHLKKAVAEI EEMCNILKTE GVTVRRPDPI DWSLKYKTPD FESTGLYSAM PRDILIVVGN EIIEAPMAWR SRFFEYRAYR SIIKDYFHRG AKWTTAPKPT MADELYNQDY PIHSVEDRHK LAAQGKFVTT EFEPCFDAAD FIRAGRDIFA QRSQVTNYLG IEWMRRHLAP DYRVHIISFK DPNPMHIDAT FNIIGPGIVL SNPDRPCHQI DLFKKAGWTI ITPPTPIIPD DHPLWMSSKW LSMNVLMLDE KRVMVDANEV PIQKMFEKLG ITTIKVNIRN ANSLGGGFHC WTCDVRRRGT LQSYLD.

AGAT, also known as glycine amidinotransferase (L-arginine:glycine amidinotransferase), GATM or transamidinase, is a 423 amino acid protein belonging to the amidinotransferase family. Encoded by a gene that maps to human chromosome 15q21.1, AGAT exists as three alternatively spliced isoforms and consists of a homodimer, with equilibrium between monomeric and dimeric forms favoring a monomer subunit structure. AGAT localizes to mitochondrial inner membranes, peripheral membranes and cytoplasm. Biallelically expressed in placenta and fetal tissues, AGAT is also expressed in brain, heart, liver, lung, salivary gland and skeletal muscle tissue, with high expression in kidney. AGAT is elevated in the myocardium during heart failure and is decreased in inter-uterine growth restriction (IUGR)-associated placenta. AGAT catalyzes biosynthesis of guanidinoacetate, the immediate precursor of creatine, which plays a vital role in energy metabolism in muscle tissues. AGAT defects are associated with arginine:glycine amidinotransferase deficiency, an autosomal recessive disorder characterized by developmental delay or regression, mental retardation, severe disturbance of expressive and cognitive speech and severe depletion of creatine/phosphocreatine in brain. AGAT may be linked to embryonic and central nervous system development and may function in heart failure response by elevating local creatine synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GATM
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene GATM
Gene Accession No. P50440, P50442, Q9D964
Gene Alias 1810003P21Rik; Agat; AI314789; AT; CCDS3; GATM; glycine amidinotransferase; glycine amidinotransferase (L-arginine:glycine amidinotransferase); glycine amidinotransferase, mitochondrial; L-arginine: glycine amidinotransferase; l-arginine:glycine amidinotransferase; testicular secretory protein Li 19; transamidinase
Gene Symbols GATM
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2628, 67092, 81660
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.