Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GATS Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144341
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144341 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144341 Supplier Invitrogen™ Supplier No. PA5144341
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GATS Polyclonal Antibody for IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen GATS
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene GATS
Gene Accession No. Q8NAP1
Gene Alias CASTOR3; GATS; GATS, stromal antigen 3 opposite strand; opposite strand transcription unit to STAG3; Putative protein CASTOR 3; putative protein GATS; STAG3 opposite strand transcript protein; STAG3OS; stromal antigen 3 opposite strand
Gene Symbols CASTOR3P, GATS
Host Species Rabbit
Immunogen Recombinant Human Putative protein CASTOR 3 protein (1-163AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 352954
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.