Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GATSL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139983
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139983 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139983 Supplier Invitrogen™ Supplier No. PA5139983
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: EQDLSVVIHT LAQEFDIYRE VGGEPVPVTR DDSSNGFPRT QHGPSPTVHP IQSPQNRFCV LTLDPET Highest antigen sequence indentity to the following orthologs: Rat - 93%, Mouse - 94%.

GATSL3 functions as an intracellular arginine sensor within the amino acid-sensing branch of the TORC1 signaling pathway. As a homodimer or a heterodimer with CASTOR2, binds and inhibits the GATOR subcomplex GATOR2 and thereby mTORC1. Binding of arginine to CASTOR1 allosterically disrupts the interaction of CASTOR1-containing dimers with GATOR2 which can in turn activate mTORC1 and the TORC1 signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GATSL3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Castor1
Gene Accession No. Q8WTX7
Gene Alias 2410008K03Rik; CASTOR1; cellular arginine sensor for mTORC1 protein 1; Cytosolic arginine sensor for mTORC1 subunit 1; Gats protein; GATS protein like 3; GATS protein-like 3; GATSL3; GATS-like protein 3; GATS-like protein 3; cytosolic arginine sensor for mTORC1 subunit 1; RGD1304774
Gene Symbols Castor1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CASTOR1. Recombinant protein control fragment (Product #RP-109325).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 652968
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.