Learn More
Invitrogen™ Gaussia luciferase Polyclonal Antibody

Description
Western blot analysis of PA1-181 detects a ~20 kDa protein in the supernatant of Gaussia luciferase-secreting cells. PA1-181 can be used for activity-independent verification of Gaussia luciferase expression.
Specifications
Specifications
| Antigen | Gaussia luciferase |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide |
| Gene Alias | Gaussia luciferase |
| Host Species | Rabbit |
| Immunogen | Purified recombinant Gaussia luciferase expressed in E. coli. |
| Purification Method | Protein G |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.