Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GBA Recombinant Rabbit Monoclonal Antibody (JM10-76)
GREENER_CHOICE

Catalog No. PIMA532591
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532591 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532591 Supplier Invitrogen™ Supplier No. MA532591
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

GBA encodes a lysosomal membrane protein that cleaves the beta-glucosidic linkage of glycosylceramide, an intermediate in glycolipid metabolism. Mutations in this gene cause Gaucher disease, a lysosomal storage disease characterized by an accumulation of glucocerebrosides. A related pseudogene is approximately 12 kb downstream of this gene on chromosome 1. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GBA
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JM10-76
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene GBA
Gene Accession No. P04062
Gene Alias acid beta glucosidase; acid beta-glucosidase; Alglucerase; betaGC; beta-GC; Beta-glucocerebrosidase; Cholesterol glucosyltransferase; Cholesteryl-beta-glucosidase; D-glucosyl-N-acylsphingosine glucohydrolase; Gba; GBA1; GBAP; GBAP1; GC; GCase; GCB; GLUC; glucocerebrosidase; glucosidase, beta, acid; glucosylceramidase; glucosylceramidase beta; glucosylceramidase beta pseudogene 1; glucosylceramidase-like protein; imiglucerase; Lysosomal acid GCase; Lysosomal acid glucosylceramidase; lysosomal glucocerebrosidase; SGTase
Gene Symbols GBA
Host Species Rabbit
Immunogen Synthetic peptide within Human GBA aa 477-534.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2629, 684536
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.