Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GBA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554955
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554955 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554955 Supplier Invitrogen™ Supplier No. PA554955
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HDTADWKDLN LKFVLQVYRD YYLTGDQNFL KDMWPVCLAV MESEMKFDKD HDGLIENGGY ADQTYDGWVT TGPSAYCGGL WLAAVAVMVQ MAALCGAQDI QDKFSSILSR GQEAYERLLW NGRYYNYDSS SRPQSRSVMS DQCA Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 95%.

Non-lysosomal glucosylceramidase that catalyzes the conversion of glucosylceramide (GlcCer) to free glucose and ceramide. Involved in sphingomyelin generation and prevention of glycolipid accumulation. May also catalyze the hydrolysis of bile acid 3-O-glucosides, however, the relevance of such activity is unclear in vivo. Plays a role in central nevous system development. Required for proper formation of motor neuron axons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GBA2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GBA2
Gene Accession No. Q9HCG7
Gene Alias 3.2.1.45; AD035; Beta-glucocerebrosidase 2; beta-glucosidase 2; bile acid; bile acid beta-glucosidase; Bile acid beta-glucosidase GBA2; Bile acid glucosyl transferase GBA2; Cholesterol glucosyltransferase GBA2; Cholesteryl-beta-glucosidase GBA2; F630034E04; GBA2; glucosidase beta 2; glucosidase, beta (bile acid) 2; glucosylceramidase 2; glucosylceramidase beta 2; Kiaa1605; NLGase; non-lysosomal glucosylceramidase; SPG46
Gene Symbols GBA2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GBA2. Recombinant protein control fragment (Product #RP-92883).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57704
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.