Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GBA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588792
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588792 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588792 Supplier Invitrogen™ Supplier No. PA588792
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: U-87MG, Mouse kidney, Mouse heart; Cellular Location: Cytoplasmic side, Endoplasmic reticulum membrane, Golgi apparatus membrane, Peripheral membrane protein.

Non-lysosomal glucosylceramidase that catalyzes the conversion of glucosylceramide (GlcCer) to free glucose and ceramide. Involved in sphingomyelin generation and prevention of glycolipid accumulation. May also catalyze the hydrolysis of bile acid 3-O-glucosides, however, the relevance of such activity is unclear in vivo. Plays a role in central nevous system development. Required for proper formation of motor neuron axons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GBA2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene GBA2
Gene Accession No. Q69ZF3, Q9HCG7
Gene Alias 3.2.1.45; AD035; Beta-glucocerebrosidase 2; beta-glucosidase 2; bile acid; bile acid beta-glucosidase; Bile acid beta-glucosidase GBA2; Bile acid glucosyl transferase GBA2; Cholesterol glucosyltransferase GBA2; Cholesteryl-beta-glucosidase GBA2; F630034E04; GBA2; glucosidase beta 2; glucosidase, beta (bile acid) 2; glucosylceramidase 2; glucosylceramidase beta 2; Kiaa1605; NLGase; non-lysosomal glucosylceramidase; SPG46
Gene Symbols GBA2
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 230101, 57704
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.