Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GBA3 Monoclonal Antibody (OTI1C7), TrueMAB™, OriGene
SDP

Catalog No. 502541430
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1430 30 μL
50-167-3643 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1430 Supplier OriGene Supplier No. TA502604S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GBA3 Monoclonal antibody specifically detects GBA3 in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen GBA3
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI1C7
Concentration 0.87 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene GBA3
Gene Accession No. Q9H227
Gene Alias CBG; CBGL1; Cytosolic beta-glucosidase; cytosolic beta-glucosidase-like protein 1; Cytosolic GCase; Cytosolic glycosylceramidase; Gba3; GLUC; Glucosidase beta acid 3; glucosidase, beta, acid 3; glucosidase, beta, acid 3 (cytosolic); Glucosylceramidase beta 3; glucosylceramidase beta 3 (gene/pseudogene); klotho-related protein; KLRP
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-150 and 370-469 of human GBA3 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57733
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.