Learn More
GBA3 Mouse anti-Human, Clone: OTI5G5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502533

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
GBA3, or cytosolic beta-glucosidase (EC 3.2.1.21), is a predomitly liver enzyme that efficiently hydrolyzes beta-D-glucoside and beta-D-galactoside, but not any known physiologic beta-glycoside, suggesting that it may be involved in detoxification of plant glycosides. GBA3 also has significant neutral glycosylceramidase activity (EC 3.2.1.62), suggesting that it may be involved in a nonlysosomal catabolic pathway of glucosylceramide metabolism.Specifications
GBA3 | |
Monoclonal | |
Unconjugated | |
GBA3 | |
CBG, CBGL1, GLUC, KLRP | |
Mouse | |
Affinity Chromatography | |
RUO | |
57733 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI5G5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9H227 | |
GBA3 | |
Human recombit protein fragment corresponding to amino acids 1-150 and 370-469 of human GBA3 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.