Learn More
Invitrogen™ GBA3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596825
Description
Immunogen sequence: MAFPAGFGWA AATAAYQVEG GWDADGKGPC VWDTFTHQGG ERVFKNQTGD VACGSYTLWE EDLKCIKQLG LTHYRFSLSW SRLLPDGTTG FINQKAIQLD KVNLQVYCAW SLLDNFEWNQ GYSSRFGLFH VDFEDPARPR VPYTSAKEYA KIIRNNGLEA HL; Positive Samples: Jurkat, HepG2, SW480, Raji, A-549, Mouse kidney; Cellular Location: Cytoplasm, cytosol.
GBA3, or cytosolic beta-glucosidase (EC 3.2.1.21), is a predominantly liver enzyme that efficiently hydrolyzes beta-D-glucoside and beta-D-galactoside, but not any known physiologic beta-glycoside, suggesting that it may be involved in detoxification of plant glycosides. GBA3 also has significant neutral glycosylceramidase activity (EC 3.2.1.62), suggesting that it may be involved in a nonlysosomal catabolic pathway of glucosylceramide metabolism.
Specifications
GBA3 | |
Polyclonal | |
Unconjugated | |
GBA3 | |
CBG; CBGL1; Cytosolic beta-glucosidase; cytosolic beta-glucosidase-like protein 1; Cytosolic GCase; Cytosolic glycosylceramidase; Gba3; GLUC; Glucosidase beta acid 3; glucosidase, beta, acid 3; glucosidase, beta, acid 3 (cytosolic); Glucosylceramidase beta 3; glucosylceramidase beta 3 (gene/pseudogene); klotho-related protein; KLRP | |
Rabbit | |
Affinity Chromatography | |
RUO | |
57733 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.41 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9H227 | |
GBA3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-162 of human GBA3 (NP_0011219041). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.