Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GBA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598985
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598985 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598985 Supplier Thermo Scientific Supplier No. PA598985
Add to Cart
Add to Cart

GBA3 Polyclonal Antibody

GBA3 Polyclonal Antibody for Western Blot, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen GBA3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.77 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GBA3
Gene Accession No. Q9H227
Gene Alias CBG; CBGL1; Cytosolic beta-glucosidase; cytosolic beta-glucosidase-like protein 1; Cytosolic GCase; Cytosolic glycosylceramidase; Gba3; GLUC; Glucosidase beta acid 3; glucosidase, beta, acid 3; glucosidase, beta, acid 3 (cytosolic); Glucosylceramidase beta 3; glucosylceramidase beta 3 (gene/pseudogene); klotho-related protein; KLRP
Gene Symbols GBA3
Host Species Rabbit
Immunogen Recombinant Human Cytosolic beta-glucosidase protein (1-162AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57733
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.